Assay Details
| Gene Symbol: SELENOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678146.3 | XP_005678203.2 | 3 - 4 | 547 | 113 | |
| GenBank mRNA | JO582151.1 | - | 2 - 3 | 945 | 113 | |
| KA338784.1 | - | 3 - 4 | 563 | 113 | ||
More Information
Set Membership: |
|

SELENOF
Genomic Map