Assay Details
Gene Symbol: SELENOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678146.3 | XP_005678203.2 | 4 - 5 | 606 | 66 | |
GenBank mRNA | JO582151.1 | - | 1 - 2 | 886 | 66 | |
KA338784.1 | - | 4 - 5 | 622 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
