Assay Details
| Gene Symbol: TUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678404.3 | XP_005678461.2 | 22 - 23 | 3873 | 73 | |
| XM_018044181.1 | XP_017899670.1 | 23 - 24 | 4012 | 73 | ||
| XM_018044190.1 | XP_017899679.1 | 22 - 23 | 3992 | 73 | ||
More Information
Set Membership: |
|

TUT4
Genomic Map