Assay Details
Gene Symbol: DKK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681333.3 | XP_005681390.1 | 1 - 2 | 943 | 71 | |
GenBank mRNA | KA341311.1 | - | 3 - 4 | 1067 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
