Assay Details
| Gene Symbol: LOC102191533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057345.1 | XP_017912834.1 | 3 - 4 | 1032 | 68 | |
| GenBank mRNA | JO577301.1 | - | 3 - 4 | 406 | 68 | |
| JT682003.1 | - | 4 - 5 | 510 | 68 | ||
More Information
Set Membership: |
|

LOC102191533
Genomic Map