Assay Details
| Gene Symbol: PRPF38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013962575.2 | XP_013818029.1 | 5 - 6 | 1058 | 73 | |
| XM_018045843.1 | XP_017901332.1 | 5 - 6 | 804 | 73 | ||
| GenBank mRNA | JO368008.2 | - | 1 - 2 | 1142 | 73 | |
| JO579267.1 | - | 1 - 2 | 1649 | 73 | ||
More Information
Set Membership: |
|

PRPF38B
Genomic Map