Assay Details
| Gene Symbol: PGM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045092.1 | XP_017900581.1 | 9 - 10 | 1848 | 62 | |
| XM_018045101.1 | XP_017900590.1 | 9 - 10 | 1663 | 62 | ||
More Information
Set Membership: |
|

PGM1
Genomic Map