Assay Details
Gene Symbol: CRPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047357.1 | XP_017902846.1 | 2 - 3 | 837 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
