Assay Details
| Gene Symbol: LOC106504024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044656.1 | XP_017900145.1 | 2575 | 76 | ||
| Gene Symbol: HTT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049859.1 | XP_017905348.1 | 8057 | 76 | ||
| XM_018049860.1 | XP_017905349.1 | 7717 | 76 | |||
| XM_018049861.1 | XP_017905350.1 | 8009 | 76 | |||
| XM_018049862.1 | XP_017905351.1 | 7925 | 76 | |||

LOC106504024
Genomic Map