Assay Details
| Gene Symbol: CRLF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051447.1 | XP_017906936.1 | 7 - 8 | 1389 | 62 | |
| GenBank mRNA | KA329109.1 | - | 7 - 8 | 1212 | 62 | |
More Information
Set Membership: |
|

CRLF1
Genomic Map