Assay Details
| Gene Symbol: C7H19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051212.1 | XP_017906701.1 | 3 - 4 | 321 | 102 | |
| XM_018051214.1 | XP_017906703.1 | 3 - 4 | 321 | 102 | ||
| GenBank mRNA | JO407982.2 | - | 2 - 3 | 220 | 102 | |
More Information
Set Membership: |
|

C7H19orf57
Genomic Map