Assay Details
Gene Symbol: C7H19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051212.1 | XP_017906701.1 | 4 - 5 | 398 | 76 | |
XM_018051214.1 | XP_017906703.1 | 4 - 5 | 398 | 76 | ||
GenBank mRNA | JO407982.2 | - | 3 - 4 | 297 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
