Assay Details
Gene Symbol: C7H19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051212.1 | XP_017906701.1 | 5 - 6 | 510 | 78 | |
XM_018051214.1 | XP_017906703.1 | 5 - 6 | 510 | 78 | ||
GenBank mRNA | JO407982.2 | - | 4 - 5 | 409 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
