Assay Details
Gene Symbol: C7H19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051214.1 | XP_017906703.1 | 6 - 7 | 1788 | 87 | |
GenBank mRNA | JO619372.2 | - | 3 - 4 | 676 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
