Assay Details
| Gene Symbol: C7H19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051214.1 | XP_017906703.1 | 8 - 9 | 1970 | 112 | |
| GenBank mRNA | JO619372.2 | - | 1 - 2 | 494 | 112 | |
More Information
Set Membership: |
|

C7H19orf57
Genomic Map