Assay Details
| Gene Symbol: DHRS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056702.1 | XP_017912191.1 | 2 - 3 | 258 | 57 | |
| XM_018056703.1 | XP_017912192.1 | 2 - 3 | 258 | 57 | ||
| GenBank mRNA | JO413777.2 | - | 8 - 9 | 1287 | 57 | |
More Information
Set Membership: |
|

DHRS12
Genomic Map