Assay Details
| Gene Symbol: LIME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057512.1 | XP_017913001.1 | 1 - 2 | 508 | 101 | |
| XM_018057513.1 | XP_017913002.1 | 1 - 2 | 508 | 101 | ||
More Information
Set Membership: |
|

LIME1
Genomic Map