Assay Details
| Gene Symbol: GPR83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969390.2 | XP_013824844.1 | 1 - 2 | 373 | 80 | |
| GenBank mRNA | KA309240.1 | - | 3 - 4 | 765 | 80 | |
More Information
Set Membership: |
|

GPR83
Genomic Map