Assay Details
| Gene Symbol: DPEP2NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013971074.2 | XP_013826528.2 | 1 - 2 | 106 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPEP2NB| Gene Symbol: DPEP2NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013971074.2 | XP_013826528.2 | 1 - 2 | 106 | 66 | |
Set Membership: |
|