Assay Details
| Gene Symbol: NPAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066431.1 | XP_017921920.1 | 10 - 11 | 1336 | 66 | |
| XM_018066432.1 | XP_017921921.1 | 10 - 11 | 1333 | 66 | ||
| XM_018066433.1 | XP_017921922.1 | 9 - 10 | 1282 | 66 | ||
| XM_018066434.1 | XP_017921923.1 | 9 - 10 | 1276 | 66 | ||
| XM_018066435.1 | XP_017921924.1 | 9 - 10 | 1282 | 66 | ||
More Information
Set Membership: |
|

NPAS3
Genomic Map