Assay Details
| Gene Symbol: SNRPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039587.1 | XP_017895076.1 | 4 - 5 | 533 | 68 | |
| XM_018039588.1 | XP_017895077.1 | 4 - 5 | 467 | 68 | ||
| GenBank mRNA | JO579096.1 | - | 1 - 2 | 285 | 68 | |
| JO634464.2 | - | 1 - 2 | 180 | 68 | ||
More Information
Set Membership: |
|

SNRPD1
Genomic Map