Assay Details
| Gene Symbol: ACAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039635.1 | XP_017895124.1 | 9 - 10 | 1211 | 71 | |
| GenBank mRNA | JO419508.2 | - | 1 - 2 | 422 | 71 | |
| JO432556.2 | - | 9 - 10 | 1302 | 71 | ||
More Information
Set Membership: |
|

ACAA2
Genomic Map