Assay Details
| Gene Symbol: NFATC2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974718.2 | XP_013830172.2 | 6 - 7 | 1106 | 70 | |
| GenBank mRNA | JO427311.1 | - | 6 - 7 | 1072 | 70 | |
More Information
Set Membership: |
|

NFATC2IP
Genomic Map