Assay Details
| Gene Symbol: LOC102191770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042322.1 | XP_017897811.1 | 15 - 16 | 1541 | 62 | |
| XM_018042323.1 | XP_017897812.1 | 15 - 16 | 1541 | 62 | ||
| XM_018042324.1 | XP_017897813.1 | 8 - 9 | 1817 | 62 | ||
More Information
Set Membership: |
|

LOC102191770
Genomic Map