Assay Details
| Gene Symbol: LOC102191770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042322.1 | XP_017897811.1 | 22 - 23 | 2404 | 104 | |
| XM_018042323.1 | XP_017897812.1 | 22 - 23 | 2404 | 104 | ||
| XM_018042324.1 | XP_017897813.1 | 15 - 16 | 2680 | 104 | ||
| GenBank mRNA | KA324377.1 | - | 22 - 23 | 2367 | 104 | |
More Information
Set Membership: |
|

LOC102191770
Genomic Map