Assay Details
| Gene Symbol: LOC102191770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042322.1 | XP_017897811.1 | 26 - 27 | 2924 | 109 | |
| XM_018042324.1 | XP_017897813.1 | 19 - 20 | 3200 | 109 | ||
| GenBank mRNA | KA324377.1 | - | 26 - 27 | 2887 | 109 | |
More Information
Set Membership: |
|

LOC102191770
Genomic Map