Assay Details
| Gene Symbol: LOC102191770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042322.1 | XP_017897811.1 | 30 - 31 | 3994 | 122 | |
| XM_018042323.1 | XP_017897812.1 | 29 - 30 | 3931 | 122 | ||
| XM_018042324.1 | XP_017897813.1 | 23 - 24 | 4270 | 122 | ||
| GenBank mRNA | JO509216.2 | - | 1 - 2 | 333 | 122 | |
| KA324377.1 | - | 30 - 31 | 3957 | 122 | ||
More Information
Set Membership: |
|

LOC102191770
Genomic Map