Assay Details
| Gene Symbol: LOC102191770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042322.1 | XP_017897811.1 | 11 - 12 | 1128 | 101 | |
| XM_018042323.1 | XP_017897812.1 | 11 - 12 | 1128 | 101 | ||
| XM_018042324.1 | XP_017897813.1 | 4 - 5 | 1404 | 101 | ||
| GenBank mRNA | KA324377.1 | - | 11 - 12 | 1097 | 101 | |
More Information
Set Membership: |
|

LOC102191770
Genomic Map