Assay Details
| Gene Symbol: MYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676482.3 | XP_005676539.1 | 4 - 5 | 762 | 68 | |
| XM_005676485.3 | XP_005676542.1 | 4 - 5 | 425 | 68 | ||
| GenBank mRNA | JO536799.2 | - | 4 - 5 | 797 | 68 | |
More Information
Set Membership: |
|

MYL1
Genomic Map