Assay Details
| Gene Symbol: PAIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050312.1 | XP_017905801.1 | 2 - 3 | 317 | 101 | |
| GenBank mRNA | JO332484.2 | - | 2 - 3 | 977 | 101 | |
More Information
Set Membership: |
|

PAIP2
Genomic Map