Assay Details
| Gene Symbol: CCNJL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050089.1 | XP_017905578.1 | 4 - 5 | 866 | 80 | |
| GenBank mRNA | JO338552.2 | - | 4 - 5 | 655 | 80 | |
More Information
Set Membership: |
|

CCNJL
Genomic Map