Assay Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052953.1 | XP_017908442.1 | 5 - 6 | 607 | 60 | |
| GenBank mRNA | JO624119.2 | - | 4 - 5 | 513 | 60 | |
More Information
Set Membership: |
|

HDDC2
Genomic Map