Assay Details
Gene Symbol: CHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685503.3 | XP_005685560.1 | 1 - 2 | 314 | 59 | |
GenBank mRNA | JO646088.2 | - | 6 - 7 | 2067 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
