Assay Details
| Gene Symbol: LARP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057346.1 | XP_017912835.1 | 13 - 14 | 4808 | 58 | |
| XM_018057347.1 | XP_017912836.1 | 15 - 16 | 2008 | 58 | ||
| GenBank mRNA | KA339171.1 | - | 3 - 4 | 667 | 58 | |
More Information
Set Membership: |
|

LARP4B
Genomic Map