Assay Details
| Gene Symbol: SLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688828.3 | XP_005688885.1 | 5 - 6 | 579 | 68 | |
| XM_013969134.2 | XP_013824588.1 | 4 - 5 | 522 | 68 | ||
| XM_013969136.2 | XP_013824590.1 | 4 - 5 | 481 | 68 | ||
| XM_018058587.1 | XP_017914076.1 | 5 - 6 | 582 | 68 | ||
More Information
Set Membership: |
|

SLA
Genomic Map