Assay Details
| Gene Symbol: LOC102172084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059551.1 | XP_017915040.1 | 30 - 31 | 4255 | 76 | |
| XM_018059552.1 | XP_017915041.1 | 30 - 31 | 4251 | 76 | ||
| XM_018059553.1 | XP_017915042.1 | 30 - 31 | 4255 | 76 | ||
| XM_018059554.1 | XP_017915043.1 | 29 - 30 | 4192 | 76 | ||
| XM_018059555.1 | XP_017915044.1 | 30 - 31 | 4132 | 76 | ||
| XM_018059556.1 | XP_017915045.1 | 28 - 29 | 4066 | 76 | ||
| XM_018059557.1 | XP_017915046.1 | 26 - 27 | 3685 | 76 | ||
| XM_018059558.1 | XP_017915047.1 | 25 - 26 | 3622 | 76 | ||
| XM_018059559.1 | XP_017915048.1 | 25 - 26 | 3577 | 76 | ||
| XM_018059560.1 | XP_017915049.1 | 24 - 25 | 3513 | 76 | ||
| GenBank mRNA | KA322253.1 | - | 7 - 8 | 2464 | 76 | |
More Information
Set Membership: |
|

LOC102172084
Genomic Map