Assay Details
| Gene Symbol: LOC102172084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059551.1 | XP_017915040.1 | 32 - 33 | 5704 | 92 | |
| XM_018059552.1 | XP_017915041.1 | 32 - 33 | 5700 | 92 | ||
| XM_018059553.1 | XP_017915042.1 | 32 - 33 | 5704 | 92 | ||
| XM_018059554.1 | XP_017915043.1 | 31 - 32 | 5641 | 92 | ||
| XM_018059555.1 | XP_017915044.1 | 32 - 33 | 5581 | 92 | ||
| XM_018059556.1 | XP_017915045.1 | 30 - 31 | 5515 | 92 | ||
| XM_018059557.1 | XP_017915046.1 | 28 - 29 | 5134 | 92 | ||
| XM_018059558.1 | XP_017915047.1 | 27 - 28 | 5071 | 92 | ||
| XM_018059559.1 | XP_017915048.1 | 27 - 28 | 5026 | 92 | ||
| XM_018059560.1 | XP_017915049.1 | 26 - 27 | 4962 | 92 | ||
| GenBank mRNA | KA322253.1 | - | 5 - 6 | 1015 | 92 | |
More Information
Set Membership: |
|

LOC102172084
Genomic Map