Assay Details
| Gene Symbol: LOC102172084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059551.1 | XP_017915040.1 | 6 - 7 | 1037 | 109 | |
| XM_018059552.1 | XP_017915041.1 | 6 - 7 | 1033 | 109 | ||
| XM_018059553.1 | XP_017915042.1 | 6 - 7 | 1037 | 109 | ||
| XM_018059554.1 | XP_017915043.1 | 6 - 7 | 1037 | 109 | ||
| XM_018059555.1 | XP_017915044.1 | 6 - 7 | 1037 | 109 | ||
| XM_018059556.1 | XP_017915045.1 | 6 - 7 | 1037 | 109 | ||
| XM_018059557.1 | XP_017915046.1 | 6 - 7 | 1037 | 109 | ||
| XM_018059558.1 | XP_017915047.1 | 6 - 7 | 1037 | 109 | ||
| XM_018059559.1 | XP_017915048.1 | 6 - 7 | 1037 | 109 | ||
| XM_018059560.1 | XP_017915049.1 | 6 - 7 | 1036 | 109 | ||
| XM_018059561.1 | XP_017915050.1 | 6 - 7 | 1031 | 109 | ||
| XM_018059562.1 | XP_017915051.1 | 6 - 7 | 1031 | 109 | ||
| XM_018059564.1 | XP_017915053.1 | 6 - 7 | 1030 | 109 | ||
| XM_018059565.1 | XP_017915054.1 | 6 - 7 | 1030 | 109 | ||
| GenBank mRNA | KA322253.1 | - | 25 - 26 | 4941 | 109 | |
More Information
Set Membership: |
|

LOC102172084
Genomic Map