Assay Details
| Gene Symbol: EXOSC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692436.3 | XP_005692493.2 | 2 - 3 | 458 | 59 | |
| GenBank mRNA | JO439265.2 | - | 4 - 5 | 762 | 59 | |
More Information
Set Membership: |
|

EXOSC5
Genomic Map