Assay Details
| Gene Symbol: PIMREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693431.3 | XP_005693488.1 | 5 - 6 | 848 | 84 | |
| GenBank mRNA | KA333978.1 | - | 1 - 2 | 204 | 84 | |
More Information
Set Membership: |
|

PIMREG
Genomic Map