Assay Details
| Gene Symbol: CCDC127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065657.1 | XP_017921146.1 | 2 - 3 | 400 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC127| Gene Symbol: CCDC127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065657.1 | XP_017921146.1 | 2 - 3 | 400 | 83 | |
Set Membership: |
|