Assay Details
| Gene Symbol: BICRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696316.3 | XP_005696373.2 | 8 - 9 | 3996 | 65 | |
| XM_018039004.1 | XP_017894493.1 | 8 - 9 | 2653 | 65 | ||
| XM_018039005.1 | XP_017894494.1 | 6 - 7 | 2476 | 65 | ||
| XM_018039006.1 | XP_017894495.1 | 8 - 9 | 2196 | 65 | ||
| XM_018039007.1 | XP_017894496.1 | 8 - 9 | 2132 | 65 | ||
| XM_018039008.1 | XP_017894497.1 | 6 - 7 | 2452 | 65 | ||
More Information
Set Membership: |
|

BICRAL
Genomic Map