Assay Details
| Gene Symbol: EEF1E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696866.2 | XP_005696923.1 | 1 - 2 | 159 | 77 | |
| GenBank mRNA | JO577887.1 | - | 3 - 4 | 699 | 77 | |
More Information
Set Membership: |
|

EEF1E1
Genomic Map