Assay Details
| Gene Symbol: BBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698468.3 | XP_005698525.1 | 3 - 4 | 537 | 83 | |
| XM_013975238.2 | XP_013830692.1 | 3 - 4 | 301 | 83 | ||
| XM_013975239.2 | XP_013830693.1 | 3 - 4 | 566 | 83 | ||
| XM_013975240.2 | XP_013830694.1 | 3 - 4 | 273 | 83 | ||
| XM_018041280.1 | XP_017896769.1 | 3 - 4 | 288 | 83 | ||
| XM_018041281.1 | XP_017896770.1 | 3 - 4 | 525 | 83 | ||
| XM_018041282.1 | XP_017896771.1 | 4 - 5 | 357 | 83 | ||
More Information
Set Membership: |
|

BBIP1
Genomic Map