Assay Details
| Gene Symbol: FRMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043448.1 | XP_017898937.1 | 10 - 11 | 1430 | 58 | |
| GenBank mRNA | KA345221.1 | - | 10 - 11 | 1462 | 58 | |
More Information
Set Membership: |
|

FRMD8
Genomic Map