Assay Details
| Gene Symbol: ATP6V0E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963581.2 | XP_013819035.2 | 2 - 3 | 373 | 103 | |
| GenBank mRNA | JO631957.2 | - | 2 - 3 | 1607 | 103 | |
| JT624308.1 | - | 2 - 3 | 187 | 103 | ||
More Information
Set Membership: |
|

ATP6V0E2
Genomic Map