Assay Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063377.1 | XP_017918866.1 | 1 - 2 | 279 | 83 | |
| XM_018063378.1 | XP_017918867.1 | 1 - 2 | 276 | 83 | ||
| GenBank mRNA | JT702686.1 | - | 5 - 6 | 1194 | 83 | |
More Information
Set Membership: |
|

THAP8
Genomic Map