Assay Details
| Gene Symbol: CPSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040201.1 | XP_017895690.1 | 4 - 5 | 578 | 80 | |
| XM_018040202.1 | XP_017895691.1 | 4 - 5 | 578 | 80 | ||
| XM_018040203.1 | XP_017895692.1 | 4 - 5 | 578 | 80 | ||
| XM_018040204.1 | XP_017895693.1 | 4 - 5 | 578 | 80 | ||
| XM_018040205.1 | XP_017895694.1 | 4 - 5 | 578 | 80 | ||
| XM_018040206.1 | XP_017895695.1 | 4 - 5 | 578 | 80 | ||
| XM_018040208.1 | XP_017895697.1 | 4 - 5 | 570 | 80 | ||
| GenBank mRNA | JO527697.3 | - | 3 - 4 | 318 | 80 | |
| JO585383.2 | - | 4 - 5 | 1147 | 80 | ||
More Information
Set Membership: |
|

CPSF4
Genomic Map