Assay Details
| Gene Symbol: PLPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041790.1 | XP_017897279.1 | 6 - 7 | 690 | 59 | |
| XM_018041791.1 | XP_017897280.1 | 7 - 8 | 755 | 59 | ||
| XM_018041792.1 | XP_017897281.1 | 6 - 7 | 648 | 59 | ||
| GenBank mRNA | KA320337.1 | - | 6 - 7 | 693 | 59 | |
More Information
Set Membership: |
|

PLPP5
Genomic Map