Assay Details
| Gene Symbol: GCSAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674914.3 | XP_005674971.2 | 3 - 4 | 314 | 72 | |
| XM_013977231.2 | XP_013832685.2 | 2 - 3 | 245 | 72 | ||
| GenBank mRNA | JO598185.2 | - | 3 - 4 | 548 | 72 | |
| JT705657.1 | - | 3 - 4 | 418 | 72 | ||
More Information
Set Membership: |
|

GCSAM
Genomic Map